sec-Butyl methyl ether 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H53427-AA
H53427-AC
H53427-AE
AAH53427-AAEA
43
USD
AAH53427-AA
AAH53427-AC
AAH53427-AE
sec-Butyl methyl ether 99%
sec-Butyl methyl ether
Purity: 99%
CAS No.: 6795-87-5
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: -30°(-22°F)
Refractive Index: 1.372
Boiling Point: 57°
UN No.: 2350
MDL No.: MFCD00190205
Formula:
C₅H₁₂O MW: 88.15 g/mol Boiling Pt: 57 °C Density: 0.742 Flash Pt: -30 °C(-22 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190205 CAS Number: 6795-87-5 UN: 2350 ADR: "3",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...