1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α-[2-(Trimethylsilyl)ethynyl]benzenemethanol

H53426-06 H53426-14
AAH53426-06EA 63.8 USD
AAH53426-06 AAH53426-14
1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol 97%
1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol

Purity: 97% CAS No.: 89530-34-7 Molecular Formula: C12H16OSi Formula Weight: 204.34 Flash Point: 110°(230°F) Refractive Index: 1.5193 MDL No.: MFCD05864336

Formula: C₁₂H₁₆OSi
Density: 0.961
Flash Pt: 110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD05864336
CAS Number: 89530-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR