N-Benzylquininium chloride 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Benzylquinine chloride, QUIBEC, (1S,2S,4S,5R)-1-Benzyl-2-((R)-hydroxy(6-methoxyquinolin-4-yl)methyl)-5-vinylquinuclidin-1-ium chloride
H53424-03
H53424-06
AAH53424-03EA
28.6
USD
AAH53424-03
AAH53424-06
AAH53424-14
N-Benzylquininium chloride 95%
N-Benzylquininium chloride
Purity: 95%
CAS No.: 67174-25-8
Molecular Formula: C27H31ClN2O2
Formula Weight: 451.01
Melting Point: 200-205° dec.
Optical Rotation: -235° (c=1.5 in water)
MDL No.: MFCD00198105
Formula:
C₂₇H₃₁ClN₂O₂ MW: 451.01 g/mol Melting Pt: 200…205 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00198105 CAS Number: 67174-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...