5-Norbornene-2-exo,3-exo-dimethanol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-exo,6-exo-Bis(hydroxymethyl)-2-norbornene, Bicyclo[2.2.1]hept-5-ene-2-exo,3-exo-dimethanol

H53422-03 H53422-06 H53422-14
AAH53422-03EA 74.29 USD
AAH53422-03 AAH53422-06 AAH53422-14
5-Norbornene-2-exo,3-exo-dimethanol 97%
5-Norbornene-2-exo,3-exo-dimethanol

Purity: 97% CAS No.: 699-95-6 Molecular Formula: C9H14O2 Formula Weight: 154.21 Flash Point: 113°(235°F) Refractive Index: 1.523 Boiling Point: 97°/20mm MDL No.: MFCD00799568

Formula: C₉H₁₄O₂
Boiling Pt: 97 °C/20 mm
Density: 1.027
Flash Pt: 113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD00799568
CAS Number: 699-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR