Methyl-4-ethynylbenzoate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl 4-ethynylbenzoate
H53419-03
H53419-06
AAH53419-03EA
138
USD
AAH53419-03
AAH53419-06
Methyl-4-ethynylbenzoate 97%
Methyl 4-ethynylbenzoate
Purity: 97%
CAS No.: 3034-86-4
Molecular Formula: C10H8O2
Formula Weight: 160.17
Melting Point: 92-94°
MDL No.: MFCD00168820
Formula:
C₁₀H₈O₂ Melting Pt: 92…94 °C Storage Temperature: Ambient |
MDL Number:
MFCD00168820 CAS Number: 3034-86-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...