1-Phenyl-1-pentyn-3-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Phenyl-1-pentyn-3-ol

H53416-03 H53416-06 H53416-14
AAH53416-03EA 32.4 USD
AAH53416-03 AAH53416-06 AAH53416-14
1-Phenyl-1-pentyn-3-ol 97%
1-Phenyl-1-pentyn-3-ol

Purity: 97% CAS No.: 27975-78-6 Molecular Formula: C11H12O Formula Weight: 160.22 Flash Point: 110°(230°F) Refractive Index: 1.559 MDL No.: MFCD00041589

Formula: C₁₁H₁₂O
MW: 160.22 g/mol
Density: 1.336
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041589
CAS Number: 27975-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR