Ethyl 3-methyl-2-oxobutyrate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 3-methyl-2-oxobutanoate
H53413-06
H53413-14
AAH53413-06EA
57.45
USD
AAH53413-06
AAH53413-14
Ethyl 3-methyl-2-oxobutyrate 97%
Ethyl-3-methyl-2-oxobutyrate
Purity: 97%
CAS No.: 20201-24-5
Molecular Formula: C7H12O3
Formula Weight: 144.17
Flash Point: 43°(109°F)
Refractive Index: 1.410
Boiling Point: 62°/11mm
Beilstein Reference: 1756668
UN No.: 3272
MDL No.: MFCD00009122
Formula:
C₇H₁₂O₃ Boiling Pt: 62 °C/11 mm Density: 0.989 Flash Pt: 43 °C (109 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009122 CAS Number: 20201-24-5 EINECS: 243-587-0 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...