2-Pentynoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pent-2-ynoic acid

H53411-03 H53411-06 H53411-14
AAH53411-03EA 33.4 USD
AAH53411-03 AAH53411-06 AAH53411-14
2-Pentynoic acid 97%
2-Pentynoic acid

Purity: 97% CAS No.: 5963-77-9 Molecular Formula: C5H6O2 Formula Weight: 98.10 Flash Point: 96°(205°F) Melting Point: 47-53° MDL No.: MFCD00014370

Formula: C₅H₆O₂
MW: 98.10 g/mol
Melting Pt: 47…53 °C
Flash Pt: 96 °C(205 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014370
CAS Number: 5963-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR