(Triisopropylsilyl)acetylene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethynyltriisopropylsilane
H53405-06
H53405-14
AAH53405-06EA
72
USD
AAH53405-06
AAH53405-14
(Triisopropylsilyl)acetylene 97%
(Triisopropylsilyl)acetylene
Purity: 97%
CAS No.: 89343-06-6
Molecular Formula: C11H22Si
Formula Weight: 182.38
Flash Point: 56°(133°F)
Refractive Index: 1.4527
Boiling Point: 50-52°/0.6mm
Beilstein Reference: 3536241
UN No.: 1993
MDL No.: MFCD00075452
Formula:
C₁₁H₂₂Si MW: 182.38 g/mol Boiling Pt: 50…52 °C/0.6 mm Density: 0.813 Flash Pt: 56 °C(133 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00075452 CAS Number: 89343-06-6 UN: 1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...