(2-Bromophenylethynyl)trimethylsilane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: ((2-Bromophenyl)ethynyl)trimethylsilane

H53402-03 H53402-06 H53402-14
AAH53402-03EA 30.3 USD
AAH53402-03 AAH53402-06 AAH53402-14
(2-Bromophenylethynyl)trimethylsilane 98%
(2-Bromophenylethynyl)trimethylsilane

Purity: 98% CAS No.: 38274-16-7 Molecular Formula: C11H13BrSi Formula Weight: 253.21 Flash Point: 113°(235°F) Physical Form: Liquid Density: 1.19 Refractive Index: 1.554 Appearance: Yellow to brown Boiling Point: 60-80°/2mm MDL No.: MFCD01321391

Formula: C₁₁H₁₃BrSi
MW: 253.21 g/mol
Boiling Pt: 60…80 °C/2 mm
Density: 1.19
Flash Pt: >113 °C(235 °F)
Storage Temperature: Ambient
MDL Number: MFCD01321391
CAS Number: 38274-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR