2-Chloro-3-formylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53399-03
H53399-06
H53399-14
AAH53399-03EA
37.3
USD
AAH53399-03
AAH53399-06
AAH53399-14
2-Chloro-3-formylpyridine 98%
2-Chloro-3-formylpyridine
Purity: 98%
CAS No.: 36404-88-3
Molecular Formula: C6H4ClNO
Formula Weight: 141.56
Flash Point:
Formula:
C₆H₄ClNO Melting Pt: 50…54 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01315308 CAS Number: 36404-88-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...