2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

H53398-03 H53398-06
AAH53398-03EA 117 USD
AAH53398-03 AAH53398-06
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride 97%
2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride

Purity: 97% CAS No.: 1171331-39-7 Molecular Formula: C4H7N2OF3.HCl Formula Weight: 192.57 MDL No.: MFCD09944516

Formula: C₄H₇N₂OF₃·HCl
MW: 192.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09944516
CAS Number: 1171331-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR