1-Chloro-5-(trimethylsilyl)-4-pentyne 97%

Supplier: Thermo Scientific Chemicals
Warning

H53393-06 H53393-14
AAH53393-06EA 45.2 USD
AAH53393-06 AAH53393-14
1-Chloro-5-(trimethylsilyl)-4-pentyne 97%
1-Chloro-5-(trimethylsilyl)-4-pentyne

Purity: 97% CAS No.: 77113-48-5 Molecular Formula: C8H15ClSi Formula Weight: 174.75 Flash Point: 65°(149°F) Refractive Index: 1.4560 Boiling Point: 75-77°/21mm MDL No.: MFCD00671353

Formula: C₈H₁₅ClSi
MW: 174.75 g/mol
Boiling Pt: 75…77 °C/21 mm
Density: 0.977
Flash Pt: 65 °C(149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671353
CAS Number: 77113-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR