1-Undecene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53387-06
H53387-14
AAH53387-06EA
63.9
USD
AAH53387-06
AAH53387-14
1-Undecene 97%
1-Undecene
Purity: 97%
CAS No.: 821-95-4
Molecular Formula: C11H22
Formula Weight: 154.30
Flash Point: 63°(145°F)
Refractive Index: 1.426
Melting Point: -49°
Boiling Point: 192-193°
Beilstein Reference: 1740044
MDL No.: MFCD00008956
Formula:
C₁₁H₂₂ MW: 154.3 g/mol Boiling Pt: 193 °C (1013 hPa) Melting Pt: -48.9 °C Density: 0.7552 g/cm³ (20 °C) Flash Pt: 63 °C(145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008956 CAS Number: 821-95-4 EINECS: 212-483-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...