1-Methylsulfonyl-1H-benzotriazole 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-(Methylsulphonyl)-1H-benzo[d][1,2,3]triazole
H53386-03
H53386-06
H53386-14
AAH53386-03EA
34.4
USD
AAH53386-03
AAH53386-06
AAH53386-14
1-Methylsulfonyl-1H-benzotriazole 97%
1-Methylsulfonyl-1H-benzotriazole
Purity: 97%
CAS No.: 37073-15-7
Molecular Formula: C7H7N3O2S
Formula Weight: 197.21
Melting Point: 106-108° dec.
MDL No.: MFCD00595638
Formula:
C₇H₇N₃O₂S MW: 197.21 g/mol Melting Pt: 106…108 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00595638 CAS Number: 37073-15-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...