4-(4-Chlorophenyl)-1-buten-4-ol 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-(4-Chlorophenyl)-3-buten-1-ol, 1-(4-Chlorophenyl)but-3-en-1-ol
H53381-03
H53381-06
AAH53381-03EA
36.6
USD
AAH53381-03
AAH53381-06
AAH53381-14
4-(4-Chlorophenyl)-1-buten-4-ol 97%
4-(4-Chlorophenyl)-1-buten-4-ol
Purity: 97%
CAS No.: 14506-33-3
Molecular Formula: C10H11ClO
Formula Weight: 182.65
Refractive Index: 1.5440
Boiling Point: 140°/2mm
MDL No.: MFCD03427246
Formula:
C₁₀H₁₁ClO MW: 182.65 g/mol Boiling Pt: 140 °C/2 mm Density: 1.137 Storage Temperature: Ambient |
MDL Number:
MFCD03427246 CAS Number: 14506-33-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...