3-(Trimethylsilyl)propiolic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(Trimethylsilyl)propynoic acid
H53380-03
H53380-06
H53380-14
AAH53380-03EA
63.5
USD
AAH53380-03
AAH53380-06
AAH53380-14
3-(Trimethylsilyl)propiolic acid 97%
3-(Trimethylsilyl)propiolic acid
Purity: 97%
CAS No.: 5683-31-8
Molecular Formula: C6H10O2Si
Formula Weight: 142.23
Flash Point: 96°(205°F)
Melting Point: 47-49°
Boiling Point: 105-110°/10mm
Beilstein Reference: 1755674
MDL No.: MFCD00190216
Formula:
C₆H₁₀O₂Si Boiling Pt: 105…110 °C/10 mm Melting Pt: 47…49 °C Flash Pt: 96 °C(205 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190216 CAS Number: 5683-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...