Boc-Arg-OH 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-2-((tert-Butoxycarbonyl)amino)-5-guanidinopentanoic acid
H53379-03
H53379-06
AAH53379-03EA
48.3
USD
AAH53379-03
AAH53379-06
AAH53379-14
Boc-Arg-OH 98%
Boc-Arg-OH
Purity: 98%
CAS No.: 13726-76-6
Molecular Formula: C11H22N4O4
Formula Weight: 274.32
Flash Point: 113°(235°F)
Beilstein Reference: 2419628
Optical Rotation: -6.5° (c=1 in acetic acid)
MDL No.: MFCD00042632
Formula:
C₁₁H₂₂N₄O₄ Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042632 CAS Number: 13726-76-6 EINECS: 237-295-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...