Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate 95%

Supplier: Thermo Scientific Chemicals

Synonyms: Bis(2-carbo-pentoxy-3,5,6-trichlorophenyl)oxalate

H53378-06 H53378-14
AAH53378-06EA 65.2 USD
AAH53378-06 AAH53378-14
Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate 95%
Bis(3,5,6-trichloro-2-n-pentyloxycarbonylphenyl) oxalate

Purity: 95% CAS No.: 75203-51-9 Molecular Formula: C26H24O8Cl6 Formula Weight: 677.19 Melting Point: 76-80° MDL No.: MFCD00012038

Formula: C₂₆H₂₄O₈Cl₆
MW: 677.19 g/mol
Melting Pt: 76…80 °C
Storage Temperature: Ambient
MDL Number: MFCD00012038
CAS Number: 75203-51-9
EINECS: 278-124-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR