1-(1-Adamantyl)ethylamine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Rimantadine Hydrochloride, alpha-Methyl-1-adamantanemethylamine Hydrochloride

H53377-06 H53377-14
AAH53377-06EA 78.9 USD
AAH53377-06 AAH53377-14
1-(1-Adamantyl)ethylamine hydrochloride 98%
1-(1-Adamantyl)ethylamine hydrochloride

Purity: 99% CAS No.: 1501-84-4 Molecular Formula: C12H21N.HCl Formula Weight: 215.77 Melting Point: 375° MDL No.: MFCD00072023

Formula: C₁₂H₂₁N
MW: 215.77 g/mol
Melting Pt: 375 °C
Storage Temperature: Ambient
MDL Number: MFCD00072023
CAS Number: 1501-84-4
Merck Index: 14,8224

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR