4-(Trimethylsilyl)-3-butyn-1-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Trimethylsilyl-3-butyn-1-ol
H53376-06
H53376-14
AAH53376-06EA
55
USD
AAH53376-06
AAH53376-14
4-(Trimethylsilyl)-3-butyn-1-ol 98%
4-(Trimethylsilyl)-3-butyn-1-ol
Purity: 98%
CAS No.: 2117-12-6
Molecular Formula: C7H14OSi
Formula Weight: 142.27
Flash Point: 60°(140°F)
Boiling Point: 97-98°/24mm
UN No.: 1993
MDL No.: MFCD00190217
Formula:
C₇H₁₄OSi MW: 142.27 g/mol Boiling Pt: 97…98 °C/24 mm Density: 0.854 Flash Pt: 60 °C (140 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190217 CAS Number: 2117-12-6 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...