1-Iodo-2-(trimethylsilyl)acetylene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H53375-03
H53375-06
H53375-14
AAH53375-03EA
41.8
USD
AAH53375-03
AAH53375-06
AAH53375-14
1-Iodo-2-(trimethylsilyl)acetylene 97%
1-Iodo-2-(trimethylsilyl)acetylene
Purity: 97%
CAS No.: 18163-47-8
Molecular Formula: C5H9ISi
Formula Weight: 224.12
Flash Point: 48°(118°F)
Refractive Index: 1.5110
Boiling Point: 130°
UN No.: 1993
MDL No.: MFCD00274201
Formula:
C₅H₉ISi Boiling Pt: 130 °C Density: 1.46 Flash Pt: 48 °C(118 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00274201 CAS Number: 18163-47-8 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...