1,3-Diphenyl-2-propyn-1-ol 90%, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,3-diphenylprop-2-yn-1-ol
H53374-03
H53374-06
AAH53374-03EA
34.8
USD
AAH53374-03
AAH53374-06
AAH53374-14
1,3-Diphenyl-2-propyn-1-ol 90%, tech.
1,3-Diphenyl-2-propyn-1-ol
Purity: 90%
CAS No.: 1817-49-8
Molecular Formula: C15H12O
Formula Weight: 208.26
Flash Point:
Formula:
C₁₅H₁₂O MW: 208.26 g/mol Density: 1.099 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD06654198 CAS Number: 1817-49-8 UN: 3082 ADR: "9",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...