2-Methyl-1-pentene 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H53373-AA
H53373-AC
H53373-AE
AAH53373-AAEA
47
USD
AAH53373-AA
AAH53373-AC
AAH53373-AE
2-Methyl-1-pentene 97%
2-Methyl-1-pentene
Purity: 97%
CAS No.: 763-29-1
Molecular Formula: C6H12
Formula Weight: 84.16
Flash Point: -27°(-17°F)
Refractive Index: 1.392
Melting Point: -136°
Boiling Point: 62°
Beilstein Reference: 1340499
UN No.: 2288
MDL No.: MFCD00009405
Formula:
C₆H₁₂ MW: 84.16 g/mol Boiling Pt: 62 °C Melting Pt: –136 °C Density: 0.7083 g/cm³ (0 °C) Flash Pt: –27 °C (–17 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009405 CAS Number: 763-29-1 EINECS: 212-108-7 UN: 3295 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...