N-(p-Toluenesulfonyl)glycine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Tosylglycine, N-p-Tosylglycine, Tos-Gly-OH

H53371-06 H53371-14
AAH53371-06EA 45.2 USD
AAH53371-06 AAH53371-14
N-(p-Toluenesulfonyl)glycine 97%
N-(p-Toluenesulfonyl)glycine

Purity: 97% CAS No.: 1080-44-0 Molecular Formula: C9H11NO4S Formula Weight: 229.25 Melting Point: 146-149° MDL No.: MFCD00045898

Formula: C₉H₁₁NO₄S
MW: 229.25 g/mol
Melting Pt: 146…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00045898
CAS Number: 1080-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR