Methyl-5-hexynoate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Methyl 5-hexynoate
H53370-06
H53370-14
AAH53370-06EA
87.1
USD
AAH53370-06
AAH53370-14
Methyl-5-hexynoate 97%
Methyl-5-hexynoate
Purity: 97%
CAS No.: 77758-51-1
Molecular Formula: C7H10O2
Formula Weight: 126.16
Refractive Index: 1.434
Boiling Point: 60-62°/10mm
MDL No.: MFCD00671366
Formula:
C₇H₁₀O₂ MW: 126.16 g/mol Boiling Pt: 60…62 °C/10 mm Storage Temperature: Ambient |
MDL Number:
MFCD00671366 CAS Number: 77758-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...