3,4-Dimethoxy-L-phenylalanine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53368-03
H53368-06
AAH53368-03EA
85.5
USD
AAH53368-03
AAH53368-06
AAH53368-14
3,4-Dimethoxy-L-phenylalanine 97%
3,4-Dimethoxy-L-phenylalanine
Purity: 97%
CAS No.: 32161-30-1
Molecular Formula: C11H15NO4
Formula Weight: 225.24
Melting Point: 254-257°
Optical Rotation: -5° (c=4 in 1M hydrochloric acid)
MDL No.: MFCD01321274
Formula:
C₁₁H₁₅NO₄ Melting Pt: 254…257 °C Storage Temperature: Ambient |
MDL Number:
MFCD01321274 CAS Number: 32161-30-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...