4-Bromo-1-butyne 97%

Supplier: Thermo Scientific Chemicals
Danger

H53367-06 H53367-14
AAH53367-06EA 104.02 USD
AAH53367-06 AAH53367-14
4-Bromo-1-butyne 97%
4-Bromo-1-butyne

Purity: 97% CAS No.: 38771-21-0 Molecular Formula: C4H5Br Formula Weight: 132.99 Storage Temperature: 2°C to 8°C Flash Point: 24°(75°F) Refractive Index: 1.481 Boiling Point: 110° UN No.: 1992 MDL No.: MFCD10000883

Formula: C₄H₅Br
MW: 132.99 g/mol
Boiling Pt: 110 °C
Density: 1.417
Flash Pt: 24 °C (75 °F)
Storage Temperature: Ambient
MDL Number: MFCD10000883
CAS Number: 38771-21-0
UN: 1992
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR