2-Chloro-5-methoxyphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Chloro-5-methoxyphenylboronic Acid, 2-Chloro-5-methoxybenzeneboronic acid
H53359-03
H53359-06
AAH53359-03EA
56.2
USD
AAH53359-03
AAH53359-06
2-Chloro-5-methoxyphenylboronic acid 98%
2-Chloro-5-methoxyphenylboronic acid
Purity: 98%
CAS No.: 89694-46-2
Molecular Formula: C7H8BClO3
Formula Weight: 186.40
Melting Point: 106-110°
MDL No.: MFCD06659858
Formula:
C₇H₈BClO₃ MW: 186.40 g/mol Melting Pt: 106…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659858 CAS Number: 89694-46-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...