2-Pyridineboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Pyridinylboronic acid, Pyridine-2-boronic acid
H53314-03
H53314-MD
AAH53314-03EA
657
USD
AAH53314-03
AAH53314-MD
2-Pyridineboronic acid 95%
2-Pyridineboronic acid
Purity: 95%
CAS No.: 197958-29-5
Molecular Formula: C5H6BNO2
Formula Weight: 122.92
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point:
Formula:
C₅H₆BNO₂ MW: 122.92 g/mol Melting Pt: >300 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00151844 CAS Number: 197958-29-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...