6-Indoleboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Indole-6-boronic acid
H53313-03
H53313-MD
AAH53313-03EA
240
USD
AAH53313-03
AAH53313-MD
6-Indoleboronic acid 98%
6-Indoleboronic acid
Purity: 98%
CAS No.: 147621-18-9
Molecular Formula: C8H8BNO2
Formula Weight: 160.97
Storage Temperature: -30°C to -10°C
Melting Point: 177-181°
MDL No.: MFCD03095176
Formula:
C₈H₈BNO₂ MW: 160.97 g/mol Melting Pt: 177…181 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03095176 CAS Number: 147621-18-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...