4-Carboxy-3-fluorophenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Borono-2-fluorobenzoic acid, 4-Carboxy-3-fluorobenzeneboronic acid

H53285-03 H53285-06
AAH53285-03EA 54.3 USD
AAH53285-03 AAH53285-06
4-Carboxy-3-fluorophenylboronic acid 98%
4-Carboxy-3-fluorophenylboronic acid

Purity: 98% CAS No.: 120153-08-4 Molecular Formula: C7H6BFO4 Formula Weight: 183.93 Melting Point: 236-240° MDL No.: MFCD01114671

Formula: C₇H₆BFO₄
Melting Pt: 236…240 °C
Storage Temperature: Ambient
MDL Number: MFCD01114671
CAS Number: 120153-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR