4-Benzyloxy-2-methylphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Benzyloxy-2-methylbenzeneboronic acid
H53266-03
H53266-06
AAH53266-03EA
118
USD
AAH53266-03
AAH53266-06
4-Benzyloxy-2-methylphenylboronic acid 98%
4-Benzyloxy-2-methylphenylboronic acid
Purity: 98%
CAS No.: 847560-49-0
Molecular Formula: C14H15BO3
Formula Weight: 242.08
Melting Point: 154-158°
MDL No.: MFCD03788404
Formula:
C₁₄H₁₅BO₃ Melting Pt: 154…158 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788404 CAS Number: 847560-49-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...