2-Phenoxyphenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenoxybenzeneboronic acid

H53264-03 H53264-06
AAH53264-03EA 38.3 USD
AAH53264-03 AAH53264-06
2-Phenoxyphenylboronic acid 98%
2-Phenoxyphenylboronic acid

Purity: 98% CAS No.: 108238-09-1 Molecular Formula: C12H11BO3 Formula Weight: 214.03 Melting Point: 116° MDL No.: MFCD01001592

Formula: C₁₂H₁₁BO₃
MW: 214.03 g/mol
Melting Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD01001592
CAS Number: 108238-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR