5-Isoquinolineboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H53230-03 H53230-MD
AAH53230-03EA 170 USD
AAH53230-03 AAH53230-MD
5-Isoquinolineboronic acid 97%
5-Isoquinolineboronic acid

Purity: 97% CAS No.: 371766-08-4 Molecular Formula: C9H8BNO2 Formula Weight: 172.98 Melting Point: 204-206° MDL No.: MFCD03839356

Formula: C₉H₈BNO₂
MW: 172.98 g/mol
Melting Pt: 204…206 °C
Storage Temperature: Ambient
MDL Number: MFCD03839356
CAS Number: 371766-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR