4-N-(Boc)aminomethylphenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-(((tert-Butoxycarbonyl)amino)methyl)phenyl)boronic acid, 4-(tert-Butoxycarbonylaminomethyl)phenylboronic acid, 4-(Aminomethyl)benzeneboronic acid, N-BOC protected

H53225-03 H53225-MD
AAH53225-03EA 251 USD
AAH53225-03 AAH53225-MD
4-N-(Boc)aminomethylphenylboronic acid 95%
4-N-(Boc)aminomethylphenylboronic acid

Purity: 97% CAS No.: 489446-42-6 Molecular Formula: C12H18BNO4 Formula Weight: 251.09 Melting Point: 204-206° MDL No.: MFCD04115637

Formula: C₁₂H₁₈BNO₄
MW: 251.09 g/mol
Melting Pt: 204…206 °C
Storage Temperature: Ambient
MDL Number: MFCD04115637
CAS Number: 489446-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR