2-(Bromomethyl)benzeneboronic acid pinacol ester 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-(2-(bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-[2-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(Bromomethyl)phenylboronic acid Pinacol ester, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl bromide

H53217-03 H53217-MD
AAH53217-03EA 112 USD
AAH53217-03 AAH53217-MD
2-(Bromomethyl)benzeneboronic acid pinacol ester 98%
2-(Bromomethyl)benzeneboronic acid pinacol ester

Purity: 98% CAS No.: 377780-72-8 Molecular Formula: C13H18BBrO2 Formula Weight: 297.00 Melting Point: 78-80° MDL No.: MFCD01632210

Formula: C₁₃H₁₈BBrO₂
MW: 297.00 g/mol
Melting Pt: 78…80 °C
Storage Temperature: Ambient
MDL Number: MFCD01632210
CAS Number: 377780-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR