3-Methyl-2-thiopheneboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Methylthiophene-2-boronic acid, (3-Methylthiophen-2-yl)boronic acid
H53201-03
H53201-MD
AAH53201-03EA
178
USD
AAH53201-03
AAH53201-MD
3-Methyl-2-thiopheneboronic acid 98%
3-Methyl-2-thiopheneboronic acid
Purity: 98%
CAS No.: 177735-09-0
Molecular Formula: C5H7BO2S
Formula Weight: 142.01
Melting Point: 192-196°
MDL No.: MFCD00236049
Formula:
C₅H₇BO₂S MW: 141.99 g/mol Melting Pt: 192…196 °C Storage Temperature: Ambient |
MDL Number:
MFCD00236049 CAS Number: 177735-09-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...