4-(1-Pyrrolidinylcarbonyl)phenylboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(1-Pyrrolidinylcarbonyl)benzeneboronic acid, 4-(Pyrrolidine-1-carbonyl)phenylboronic acid, (4-(Pyrrolidine-1-carbonyl)phenyl)boronic acid

H53186-03 H53186-06
AAH53186-03EA 67.1 USD
AAH53186-03 AAH53186-06
4-(1-Pyrrolidinylcarbonyl)phenylboronic acid 96%
4-(1-Pyrrolidinylcarbonyl)phenylboronic acid

Purity: 96% CAS No.: 389621-81-2 Molecular Formula: C11H14BNO3 Formula Weight: 219.05 Melting Point: 136-140° MDL No.: MFCD03411950

Formula: C₁₁H₁₄BNO₃
MW: 219.05 g/mol
Melting Pt: 136…140 °C
Storage Temperature: Ambient
MDL Number: MFCD03411950
CAS Number: 389621-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR