3-(Methylcarbamoyl)phenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-(N-Methylaminocarbonyl)phenylboronic acid, 3-(Methylcarbamoyl)benzeneboronic acid, (3-(Methylcarbamoyl)phenyl)boronic acid
H53141-03
H53141-MD
AAH53141-03EA
265
USD
AAH53141-03
AAH53141-MD
3-(Methylcarbamoyl)phenylboronic acid 98%
3-(Methylcarbamoyl)phenylboronic acid
Purity: 98%
CAS No.: 832695-88-2
Molecular Formula: C8H10BNO3
Formula Weight: 178.98
Melting Point: 114-124°
MDL No.: MFCD04038918
Formula:
C₈H₁₀BNO₃ Melting Pt: 114…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD04038918 CAS Number: 832695-88-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...