3-Chloro-4-(trifluoromethyl)phenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Chloro-4-(trifluoromethyl)benzeneboronic acid
H53120-03
H53120-MD
AAH53120-03EA
117
USD
AAH53120-03
AAH53120-MD
3-Chloro-4-(trifluoromethyl)phenylboronic acid 97%
3-Chloro-4-(trifluoromethyl)phenylboronic acid
Purity: 97%
CAS No.: 847756-88-1
Molecular Formula: C7H5BClF3O2
Formula Weight: 224.38
Melting Point: 206-212°
MDL No.: MFCD03788417
Formula:
C₇H₅BClF₃O₂ MW: 224.38 g/mol Melting Pt: 206…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788417 CAS Number: 847756-88-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...