(3-Fluoro-4-hydroxyphenyl)boronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Fluoro-4-hydroxyphenylboronic acid, 3-Fluoro-4-hydroxybenzeneboronic acid
H53110-03
H53110-MD
AAH53110-03EA
284
USD
AAH53110-03
AAH53110-MD
(3-Fluoro-4-hydroxyphenyl)boronic acid 97%
(3-Fluoro-4-hydroxyphenyl)boronic acid
Purity: 97%
CAS No.: 182344-14-5
Molecular Formula: C6H6BFO3
Formula Weight: 155.92
Melting Point: 252-256°
MDL No.: MFCD06659838
Formula:
C₅H₅BFNO₂ MW: 155.92 g/mol Melting Pt: 252…256 °C Storage Temperature: Ambient |
MDL Number:
MFCD06659838 CAS Number: 182344-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...