3-Carbamoyl-4-fluorobenzeneboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Aminocarbonyl)-4-fluorophenylboronic acid, (3-Carbamoyl-4-fluorophenyl)boronic acid

H53102-03 H53102-MD
AAH53102-03EA 228 USD
AAH53102-03 AAH53102-MD
3-Carbamoyl-4-fluorobenzeneboronic acid 97%
3-Carbamoyl-4-fluorobenzeneboronic acid

Purity: 97% CAS No.: 874219-34-8 Molecular Formula: C7H7BFNO3 Formula Weight: 182.95 Melting Point: 257-259° MDL No.: MFCD08235053

Formula: C₇H₇BFNO₃
MW: 182.95 g/mol
Melting Pt: 257…259 °C
Storage Temperature: Ambient
MDL Number: MFCD08235053
CAS Number: 874219-34-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR