4-Isocyanatobenzeneboronic acid pinacol ester 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H53003-03
H53003-MD
AAH53003-03EA
250
USD
AAH53003-03
AAH53003-MD
4-Isocyanatobenzeneboronic acid pinacol ester 95%
4-Isocyanatobenzeneboronic acid pinacol ester
Purity: 95%
CAS No.: 380430-64-8
Molecular Formula: C13H16BNO3
Formula Weight: 245.08
Melting Point: 162-168°
MDL No.: MFCD03411942
Formula:
C₁₃H₁₆BNO₃ Melting Pt: 162…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411942 CAS Number: 380430-64-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...