4-Methyl-1-naphthaleneboronic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H52998-03
H52998-06
AAH52998-03EA
36.5
USD
AAH52998-03
AAH52998-06
4-Methyl-1-naphthaleneboronic acid 96%
4-Methyl-1-naphthaleneboronic acid
Purity: 96%
CAS No.: 103986-53-4
Molecular Formula: C11H11BO2
Formula Weight: 186.02
Melting Point: 239-240°
MDL No.: MFCD01632204
Formula:
C₁₁H₁₁BO₂ MW: 186.02 g/mol Melting Pt: 239…240 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632204 CAS Number: 103986-53-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...