(4-Acetamidophenyl)boronic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Acetamidophenylboronic acid, 4-Acetamidobenzeneboronic acid, 4-Acetylaminophenylboronic acid
H52987-03
H52987-06
AAH52987-03EA
92.6
USD
AAH52987-03
AAH52987-06
(4-Acetamidophenyl)boronic acid 96%
(4-Acetamidophenyl)boronic acid
Purity: 96%
CAS No.: 101251-09-6
Molecular Formula: C8H10BNO3
Formula Weight: 178.98
Melting Point: 215-220°
MDL No.: MFCD02179451
Formula:
C₈H₁₀BNO₃ MW: 178.98 g/mol Melting Pt: 215…220 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179451 CAS Number: 101251-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...