3,5-Dimethoxyphenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3,5-Dimethoxybenzeneboronic acid
H52965-03
H52965-06
AAH52965-03EA
55.6
USD
AAH52965-03
AAH52965-06
3,5-Dimethoxyphenylboronic acid 98%
3,5-Dimethoxyphenylboronic acid
Purity: 98%
CAS No.: 192182-54-0
Molecular Formula: C8H11BO4
Formula Weight: 181.98
Melting Point: 205-207°
MDL No.: MFCD03095127
Formula:
C₈H₁₁BO₄ MW: 181.98 g/mol Melting Pt: 205…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095127 CAS Number: 192182-54-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...