5-Bromo-2-hydroxybenzeneboronic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H52937-03
H52937-MD
AAH52937-03EA
214
USD
AAH52937-03
AAH52937-MD
5-Bromo-2-hydroxybenzeneboronic acid 96%
5-Bromo-2-hydroxybenzeneboronic acid
Purity: 97%
CAS No.: 89598-97-0
Molecular Formula: C6H6BBrO3
Formula Weight: 216.83
Physical Form: Solid
Appearance: White
Melting Point: 196-200°
MDL No.: MFCD00093408
Formula:
C₆H₆BBrO₃ Melting Pt: 196…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD00093408 CAS Number: 89598-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...