4-(Isopropylcarbamoyl)phenylboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(N-Isopropylaminocarbonyl)phenylboronic acid, (4-(Isopropylcarbamoyl)phenyl)boronic acid, 4-(Isopropylcarbamoyl)benzeneboronic acid
H52925-03
H52925-MD
AAH52925-03EA
181
USD
AAH52925-03
AAH52925-MD
4-(Isopropylcarbamoyl)phenylboronic acid 95%
4-(Isopropylcarbamoyl)phenylboronic acid
Purity: 95%
CAS No.: 397843-67-3
Molecular Formula: C10H14BNO3
Formula Weight: 207.04
Melting Point: 162-167°
MDL No.: MFCD04039353
Formula:
C₁₀H₁₄BNO₃ MW: 207.04 g/mol Melting Pt: 162…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039353 CAS Number: 397843-67-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...