3-(Methylsulfonyl)phenylboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-(Methanesulphonyl)phenylboronic acid, 3-(Methylsulphonyl)benzeneboronic acid, (3-(Methylsulphonyl)phenyl)boronic acid

H52916-03 H52916-06
AAH52916-03EA 120 USD
AAH52916-03 AAH52916-06
3-(Methylsulfonyl)phenylboronic acid 98%
3-(Methylsulphonyl)phenylboronic acid

Purity: 98% CAS No.: 373384-18-0 Molecular Formula: C7H9BO4S Formula Weight: 200.02 Melting Point: 102-110° MDL No.: MFCD03092935

Formula: C₇H₉BO₄S
MW: 200.02 g/mol
Melting Pt: 102…110 °C
Storage Temperature: Ambient
MDL Number: MFCD03092935
CAS Number: 373384-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR